missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate, anhydrous, 98%

Catalog No. p-7021587
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1398630 250 g
AAA139860B 1000 g
AAA139860I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1398630 Supplier Thermo Scientific Chemicals Supplier No. A1398630
Only null left

Copper(II) sulfate, 98%, CuO4S, CAS Number-7758-98-7, copper 2+ sulfate, copper ii sulfate, blue stone, copper monosulfate, copper sulphate, trinagle, copper sulfate, hylinec, cupric sulfate anhydrous, copper ii sulfate, anhydrous, 1000g, CHEBI:23414, 3.6, Hazard Class: 9; Packaging Group: III | CAS: 7758-98-7 | CuO4S | 159.602 g/mol

Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling′s solution and Benedict′s solution to test for reducing sugars. Also, used to test protein in Biuret test.

Solubility
Soluble in water and methanol. Insoluble in ethanol.

Notes
Incompatible with strong bases, metals, alkali metals and powdered metals.

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Melting Point >560°C
Quantity 250 g
Linear Formula CuSO4
UN Number UN3288
Merck Index 14,2653
Solubility Information Soluble in water and methanol. Insoluble in ethanol.
Formula Weight 159.6
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 3.6
Chemical Name or Material Copper(II) sulfate, anhydrous
Show More Show Less
missing translation for 'healthHazard1' H302-H315-H319
missing translation for 'healthHazard2' GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
missing translation for 'healthHazard3' P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-847-6
RTECSNumber GL8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only