missing translation for 'onlineSavingsMsg'
Learn More

Copper (II) Sulfate, 1%, Fisher Chemical™

Catalog No. SC9201500
Change view
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SC9201500 500 mL Natural Poly Bottle
SC9201250 250 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SC9201500 Supplier Ricca Chemical Company Supplier No. SC9201500

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Percent Purity 1%
Grade Laboratory
Packaging Natural Poly Bottle
Methods For AOAC Method 960.27
Quantity 500 mL
Chemical Name or Material Copper (II) Sulfate, 1%