missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, AAS standard solution, Specpure™ Cu 1000μg/mL
Description
Copper, AAS standard solution is used to determine copper in samples of water and water-suspendedsediment with a specific conductance not greater than 10,000 μS/cm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Vapor Pressure | 17mm Hg at 20°C |
| Physical Form | Liquid |
| Color | Colorless |
| Solubility | Miscible with water |
| Quantity | 100 mL |
| Grade | AAS Standard |
| Concentration or Composition (by Analyte or Components) | Nitric acid: 5%; Copper: 0.1%; Water: 94.9% |
| Chemical Name or Material | Copper |
Safety and Handling
| Hazard Category | H290-H315-H318 |
| Precautionary Statement | P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390 |
| DOTInformation | DOT Class: 8, Packaging Group: III |
| EINECSNumber | 231-714-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only