missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ QuickTask™ Flat Hinged Mop Head Frames

Contec™ QuickTask™ Flat Hinged Mop Head Frames are durable, lightweight and exclusively designed for the Contec™ QuickTask™ Mop System.
$69.91 - $764.11
| Catalog Number | Mfr. No. | Material | Dimensions | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Dimensions | Price | Quantity & Availability | ||||
19168796
![]() |
Contec™
2740 |
Polypropylene | 41 x 13 cm (16 x 5 in.) |
Each for $69.91
|
|
||||
19161025
![]() |
Contec™
CTC2906 |
Stainless Steel | 40 x 10 cm (15.5 x 3.75 in.) |
Each for $764.11
|
|
||||
19161026
![]() |
Contec™
2699P |
Polypropylene | 40 x 10 cm (15.5 x 3.75 in.) |
Each for $235.97
|
|
||||
Description
- Available in stainless steel or blue polypropylene
- Autoclavable
- Compatible with most common disinfectant solutions and solvents
- The QuickTask™ system consists of a variety of knitted polyester, polyester/cellulose, microfiber/foam, and microfiber flat mop heads that are easily installed onto a hinged mop head frame and handle assembly
- The QuickTask™ system is ideally suited for cleaning floors, including perforated cleanroom floors and grates
Specifications
| Non-sterile | |
| Autoclavable |