Learn More
Thermo Scientific™ Consumables for KingFisher™ mL Systems
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
$558.80 - $1,966.80
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
22387024
|
Thermo Scientific™
97002111 |
800 pieces |
Pack of 800 Each for $1,966.80
|
|
|||||
|
22387025
|
Thermo Scientific™
97002121 |
20 x 45 pieces |
Pack of 900 Each for $1,001.00
|
|
|||||
22387026
|
Thermo Scientific™
97002141 |
For 240 samples |
Each for $558.80
|
|
|||||
Description
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
- One sample processing uses one tube strip containing five tubes.
- One tip comb with five tips is used for processing five samples at a time.
- One sample processing uses one tube strip containing five tubes.
- One tip comb with five tips is used for processing five samples at a time.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.