Learn More
Thermo Scientific™ Consumables for KingFisher™ mL Systems
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
Supplier:  Thermo Scientific™ 97002121
 View more versions of this product
Catalog No. 22387025
Only null left
Description
Description
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
The KingFisher mL instrument is designed for a maximum of 15 tube strips, which are compatible with the specially desinged tip comb.
  • One sample processing uses one tube strip containing five tubes.
  • One tip comb with five tips is used for processing five samples at a time.
Specifications
Specifications
20 x 45 pieces
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.