missing translation for 'onlineSavingsMsg'
Learn More

Congo Red, Spectrum™ Chemical

$390.71 - $2,562.96

Chemical Identifiers

CAS 573-58-0
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18609909
View Documents Promotion Details
N/A 10 g Amber Glass Bottle
Each for $390.71
Only null left
 
18609911
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $661.61
Only null left
 
18609910
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $1,465.93
Only null left
 
18609912
View Documents Promotion Details
N/A 500 g Amber Glass Bottle
Each for $2,562.96
Only null left
 
Description

Description

Specifications

Chemical Identifiers

573-58-0

Specifications

573-58-0
Reagent
Congo Red
1
Red/Brown to Dark Red
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.