missing translation for 'onlineSavingsMsg'
Learn More

Congo Red, Spectrum™ Chemical

Catalog No. 18609909
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609909 10 g Amber Glass Bottle
18609911 25 g Amber Glass Bottle
18609910 100 g Amber Glass Bottle
18609912 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609909 Supplier Spectrum Chemical Mfg Cor Supplier No. CO12810GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 573-58-0

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Red/Brown to Dark Red
Quantity 10 g
Chemical Name or Material Congo Red

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.