missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Congo Red, indicator grade

CAS: 573-58-0 | C32H22N6Na2O6S2 | 696.664 g/mol

$108.29 - $976.67

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
MDL Number MFCD00004028
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym C.I. 22120
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAB2431014
View Documents Promotion Details
Thermo Scientific Chemicals
B2431014
25 g
Each for $108.29
Only null left
 
AAB2431022
View Documents Promotion Details
Thermo Scientific Chemicals
B2431022
100 g
Each for $294.22
Only null left
 
AAB2431036
View Documents Promotion Details
Thermo Scientific Chemicals
B2431036
500 g
Each for $976.67
Only null left
 
Description

Description

Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

Solubility
Soluble in water 25 g/L.

Notes
Store away from oxidizing agents, acids, reducing agents.
Specifications

Chemical Identifiers

573-58-0
696.664
IQFVPQOLBLOTPF-UHFFFAOYSA-L
11313
disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
C32H22N6Na2O6S2
MFCD00004028
C.I. 22120
CHEBI:34653
C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

>360°C
C32H22N6Na2O6S2
3894858
C.I. 22120
IQFVPQOLBLOTPF-UHFFFAOYSA-L
disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
11313
696.67
Odorless
573-58-0
MFCD00004028
14,2498
Soluble in water 25g/L.
C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
696.664
CHEBI:34653
Indicator
Congo Red
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H361
May cause cancer.
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H350-H361d

EINECSNumber : 209-358-4

RTECSNumber : QK1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only