missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Congo Red, indicator grade

CAS: 573-58-0 | C32H22N6Na2O6S2 | 696.664 g/mol

Supplier:  Thermo Scientific Chemicals B2431022

 View more versions of this product

Catalog No. AAB2431022


Only null left

Description

Description

Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

Solubility
Soluble in water 25 g/L.

Notes
Store away from oxidizing agents, acids, reducing agents.
Specifications

Specifications

Congo Red
573-58-0
MFCD00004028
14,2498
Soluble in water 25g/L.
C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
696.664
CHEBI:34653
Indicator
100 g
>360°C
C32H22N6Na2O6S2
3894858
C.I. 22120
IQFVPQOLBLOTPF-UHFFFAOYSA-L
disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
11313
696.67
Odorless

Chemical Identifiers

573-58-0
696.664
IQFVPQOLBLOTPF-UHFFFAOYSA-L
11313
disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
C32H22N6Na2O6S2
MFCD00004028
C.I. 22120
CHEBI:34653
C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H361
May cause cancer.
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H350-H361d

EINECSNumber : 209-358-4

RTECSNumber : QK1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only