missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 50 μS/cm at 25°C (23.6 ppm TDS as NaCl), Ricca Chemical
$115.16 - $395.97
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
223610-16
|
Ricca Chemical Company
22361016 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
|
223610-32
|
Ricca Chemical Company
22361032 |
1 L | Natural Poly Bottle |
N/A
|
|
||||
|
223610-1
|
Ricca Chemical Company
2236101 |
4 L | Natural Poly Bottle |
N/A
|
|
||||
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium chloride |
Specifications
| Natural Poly Bottle | |
| 23.6ppm as NaCl | |
| ClNa | |
| Colorless | |
| Conductivity Standard | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 500 mL | |
| CHEBI:26710 |
| 7647-14-5,7732-18-5 | |
| Liquid | |
| MFCD00003477 | |
| 100°C | |
| Conductivity Standard | |
| [Na+].[Cl-] | |
| 58.44 | |
| 5234 | |
| Conductivity/TDS Standard |