missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 7000 μS/cm at 25°C (3718 ppm TDS as NaCl), Ricca Chemical

$118.40 - $439.67

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
CAS 7647-14-5,7732-18-5
CAS Min % 98.0,0.36
Concentration or Composition Notes 3718ppm as NaCl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
224570-16
View Documents
Ricca Chemical Company
22457016
500 mL
Each for $118.40
Only null left
 
224570-32
View Documents
Ricca Chemical Company
22457032
1 L
Each for $195.00
Only null left
 
224570-1
View Documents
Ricca Chemical Company
2245701
4 L
Each for $439.67
Only null left
 
Description

Description

Specifications

Specifications

Conductivity/TDS Standard
7647-14-5,7732-18-5
3718ppm as NaCl
Liquid
100°C
Conductivity Standard
Natural Poly Bottle
98.0,0.36
100.0,0.38
Colorless
Conductivity Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'