missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 7000 μS/cm at 25°C (3718 ppm TDS as NaCl), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Conductivity/TDS Standard |
| Packaging | Natural Poly Bottle |
| CAS | 7647-14-5, 7732-18-5 |
| Concentration or Composition Notes | 3718ppm as NaCl |
| CAS Min % | 98.0, 0.36 |
| CAS Max % | 100.0, 0.38 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Solution Type | Conductivity Standard |
| Show More |