missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C, Ricca Chemical

5400 ppm TDS as NaCl

$128.13 - $474.88

Specifications

Chemical Name or Material Conductivity/TDS Standard
Name Note 10mS/cm ((10, 000μmho/cm) at 25°C, 5400ppm as NaCl
CAS 7647-14-5,7732-18-5
CAS Min % 97.0,0.53
CAS Max % 100.0,0.55
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
2246100P
View Documents
Ricca Chemical Company
2246100P
100 mL Pack
Each for $200.63
Only null left
 
2246-16
View Documents
Ricca Chemical Company
224616
500 mL Natural Poly Bottle
Each for $128.13
Only null left
 
2246-32
View Documents
Ricca Chemical Company
224632
1 L Natural Poly Bottle
Each for $197.78
Only null left
 
22461
View Documents
Ricca Chemical Company
22461
4 L Natural Poly Bottle
Each for $474.88
Only null left
 
Description

Description

Specifications

Specifications

Conductivity/TDS Standard
7647-14-5,7732-18-5
100.0,0.55
Colorless
Conductivity Standard
5400ppm as NaCl
10mS/cm ((10, 000μmho/cm) at 25°C, 5400ppm as NaCl
97.0,0.53
Liquid
100°C
Conductivity Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'