missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C, Ricca Chemical

Catalog No. 224632
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
224632 1 L Natural Poly Bottle
2246100P 100 mL Pack
224616 500 mL Natural Poly Bottle
22461 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 224632 Supplier Ricca Chemical Company Supplier No. 224632
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5400 ppm TDS as NaCl

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Natural Poly Bottle
Name Note 10mS/cm ((10, 000μmho/cm) at 25°C, 5400ppm as NaCl
CAS 7647-14-5, 7732-18-5
CAS Min % 97.0, 0.53
CAS Max % 100.0, 0.55
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Concentration or Composition (by Analyte or Components) 5400ppm as NaCl
Show More Show Less