missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Con A Sepharose™ 4B Medium

Catalog No. p-3667108
Change view
Click to view available options
Quantity:
5 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000069 5 mL
45000068 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000069 Supplier Cytiva Supplier No. 17044003
Only null left

An affinity chromatography media for capture of glycosylated biomolecules including glycoproteins and glycolipids

Highly suited for isolation of cell surface glycoproteins from detergent-solubilized membranes.

  • Con A coupled to Sepharose, in a 4% agarose matrix via cyanogen bromide activation
  • Binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
  • Group-specific adsorbent for molecules containing sugars
  • Concentration of immobilized Con A, approx. 13mg/mL medium; binding capacity for porcine thyroglobulin, approx. 30mg/mL medium
  • Stable in the pH range 4 to 9, and in the presence of all commonly used buffers
  • Average particle size: 90μm
  • Delivered in 0.1M acetate buffer, pH 6.0, containing 1M NaCl, 1mM CaCl2, 1mM MgCl2, 1mM MnCl2, and 20% ethanol

Specifications

Binding Capacity 30mg/mL
Ligand Concentration 13mg Con A/mL Drained Medium
Matrix 4% Agarose
Particle Size 90 μm
pH 4 to 9
Quantity 5 mL
Content And Storage 2°C to 8°C
For Use With (Application) For separation and purification of Glycoproteins, Polysaccharides and Glycolipids, isolates cell surface Glycoproteins from detergent-solubilized membranes
Ligand Concanavalin A