missing translation for 'onlineSavingsMsg'
Learn More

Complex Cyanide Standard, 1000μg/mL in water, Ultra Scientific

Catalog No. USICC009
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICC009 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICC009 Supplier Brands Supplier No. ICC009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13746-66-2 , 7732-18-5
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

CAS Max % 99.78, 0.21
Analyte or Component Names Water,Potassium ferricyanide
Element Potassium Ferricyanide
Solvent or Matrix Water
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Complex Cyanide Standard