Learn More
OHAUS™ Compass™ Cx Compact Scale
Designed for energy efficient weighing in labs, classrooms or the field.
Supplier:  OHAUS™ 30428201DUP
 View more versions of this product
Catalog No. 01922219
Only null left
Explore available promotions
Description
Description
  • Up to 1,000 hours of continuous operation using standard alkaline batteries
  • Large backlit display
  • Stackable design.
Specifications
Specifications
1200 g
0.1 g
2 sec
201 mm
41 mm
Backlit LCD
Stainless steel pan, AA batteries
Compact Scale
135 mm
0.51 kg
Product Suggestions
Product Suggestions