missing translation for 'onlineSavingsMsg'
Learn More

Promega Column Wash Solution (CWA) for Wizard™ SV 96 Purification Systems DFS Item

Column Wash Solution (CWA) is a component of the Wizard SV and Wizard SV 96 Genomic, Wizard Plus SV Minipreps and Wizard SV 96 and SV 9600 Plasmid DNA Purification Systems.

$113.24 - $274.07

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Purification system
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Product Type Price Quantity  
Catalog Number Mfr. No. Quantity Product Type Price Quantity  
PR-A1318 DFS Item
View Documents
Promega
A1318
370 mL SV 96 Wash Solution
Each for $274.07
Only null left
 
PR-A1311 DFS Item
View Documents
Promega
A1311
185 mL SV Wash Solution
Each for $113.24
Only null left
 
Description

Description

  • Wash Solution used for Automated DNA Purification Protocols
  • Component of the Wizard(R) SV, Wizard(R) SV 96 Genomic, and Wizard SV 96 and SV 9600 Plasmid DNA Purification Systems available as a standalone item
Specifications

Specifications

15°C to 30°C
Purification system
Videos
SDS
Documents

Documents