Learn More
MilliporeSigma™ Supelco™ column Blank Kit for HPLC
Each column blank kit includes a precision-bore polished 316 stainless steel column blank, two zero dead volume external reducing unions (internal unions with 10 mm I.D. blanks) with nuts and ferrules for 1/16 in. O.D. tubing, and two 2 μm frits.
Supplier:  MilliporeSigma™ Supelco™ 58217
 View more versions of this product
Catalog No. 111010261
Only null left
Description
Description
Specifications
Chemical Identifiers
316 L Stainless Steel
Specifications
316 L Stainless Steel
I.D. O.D. L 10 mm 1/2 in. 25 cm
Product Suggestions
Product Suggestions