missing translation for 'onlineSavingsMsg'
Learn More

Color-Coded Spring Scales

Catalog No. p-4133082
Change view
Click to view available options
Capacity:
1.1 lb., 500 g
11 lb., 5 kg
Color:
Green
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Color
S02322 1.1 lb., 500 g Green
S02326 11 lb., 5 kg Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02322 Supplier Customer Channels Group (Ccg) Supplier No. 39610
Only null left

Clear plastic tube allows students to see the action of the spring.

Series of eight color-coded tubular spring scales can be used for both force and mass measurements.

  • Calibrated in both Newtons and grams with a beveled top for use on an inclined plane
  • Incorporates a unique zero adjustment

Order Info

Available as a set of six colors (Cat. No. S02321A).

Specifications

Color Green
Grade 4+
Capacity 1.1 lb., 500 g
Type Spring Scale