missing translation for 'onlineSavingsMsg'
Learn More

Colchicine, 98%

Microtubule disrupting agent, and induces apoptosis in human lymphoma cells | CAS: 64-86-8 | C22H25NO6 | 399.44 g/mol

Supplier:  Thermo Scientific Chemicals J6107206

 View more versions of this product

Catalog No. AAJ6107206


Only null left
Explore available promotions

Description

Description

Colchicine is most commonly used to treat gout. It is an inhibitor of microtubule polymerization which blocks chromosome segregation during meiosis. Consequently, it is used to induce polyploidy (tetraploid) in plant cells. It is an antimitotic agent that disrupts microtubules by binding to tubulin and preventing its polymerization. Stimulates the intrinsic GTPase activity of tubulin. Induces apoptosis in several normal and tumor cell lines and activates the JNK/SAPK signaling pathway.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Colchicine is most commonly used to treat gout. It is an inhibitor of microtubule polymerization which blocks chromosome segregation during meiosis. Consequently, it is used to induce polyploidy (tetraploid) in plant cells. It is an antimitotic agent that disrupts microtubules by binding to tubulin and preventing its polymerization. Stimulates the intrinsic GTPase activity of tubulin. Induces apoptosis in several normal and tumor cell lines and activates the JNK/SAPK signaling pathway.

Solubility
Soluble in ethanol (10 mg/ml), water (45 g/L), chloroform, DMSO and methanol.

Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

Colchicine
∽150°C (decomposition)
C22H25NO6
5 g
2228813
14,2471
Soluble in ethanol (10mg/ml),water (45g/L),chloroform,DMSO and methanol.
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1
399.44
399.44
Powder
64-86-8
White to Yellow
MFCD00078484,MFCD00078484
UN1544
Light sensitive
colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine
IAKHMKGGTNLKSZ-INIZCTEOSA-N
2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
45038708
98%

Chemical Identifiers

64-86-8
399.44
IAKHMKGGTNLKSZ-INIZCTEOSA-N
45038708
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1
C22H25NO6
MFCD00078484,MFCD00078484
colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine
2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H340
Fatal if swallowed.
May cause genetic defects.

P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c

H300-H340

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.

EINECSNumber : 200-598-5

RTECSNumber : GH0700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only