missing translation for 'onlineSavingsMsg'
Learn More

COD Standard, 300ppm (1mL = 0.3mg COD) (Potassium Acid Phthalate in Water), Certified, 0.300mg/mL ±0.003, LabChem™

Supplier:  LabChem LC132401

Catalog No. LC132401


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

COD Standard
Poly Bottle
99.67,0.03,0.3
Liquid
C8H5KO4
KHC8H4O4
Soluble in water
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
23676735
Certified
300ppm (1mL = 0.3mg COD) (Potassium Acid Phthalate in Water)
877-24-7 , 7664-38-2 , 7732-18-5
Passes Test
as COD
0.300 mg/mL ±0.003
Colorless
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
500 mL
204.22

Chemical Identifiers

877-24-7
204.222
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions