missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Sulfate, Heptahydrate, Crystal, 97%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C132512KG
Specifications
| 10026-24-1 | |
| 12 kg | |
| MFCD00149658 | |
| O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O | |
| 281.09 | |
| Reagent |
| 1 | |
| CoH14O11S | |
| MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
| λ²-cobalt(2+) heptahydrate sulfate | |
| 97% | |
| Poly Pail |
Chemical Identifiers
| 10026-24-1 | |
| 281.09 | |
| MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
| O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
| CoH14O11S | |
| MFCD00149658 | |
| λ²-cobalt(2+) heptahydrate sulfate |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.