accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2OCoOSOOO

CAS RN 10026-24-1

IUPAC Name: λ²-cobalt(2+) heptahydrate sulfate
Synonyms: cobalt(II) sulfate
Molecular Weight (g/mol): 281.09
Molecular Formula: CoH14O11S
InChi Key: MEYVLGVRTYSQHI-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
Grade 
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (6)
  • (3)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (9)
  • (10)
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (6)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)