missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Sulfate, Heptahydrate, Crystal, 97%, Spectrum™ Chemical

Catalog No. p-7165340
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18609865 25 g Poly Bottle
18609866 125 g Poly Bottle
18609867 500 g Poly Bottle
18609868 2.5 kg Poly Bottle
18609869 12 kg Poly Pail
18609870 50 kg Fiber Drum
6 options

Catalog No. 18609865

Supplier: Spectrum Chemical Mfg Cor C132525GM

Only null left

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.