missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
CAS: 10141-05-6 | CoN2O6 | 182.941 g/mol
$57.86 - $57.86
Specifications
| Chemical Name or Material | Cobalt standard solution |
|---|---|
| Name Note | 1 mg/ml Co in 2% HNO3 |
| Packaging | Plastic bottle |
| CAS | 10141-05-6 , 7732-18-5 |
| Physical Form | Solution |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC195921000
|
Thermo Scientific Chemicals
195921000 |
100 mL |
Each for $57.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
| Cobalt standard solution | |
| Plastic bottle | |
| Solution | |
| 990 to 1010 μg/mL of Co(2+) in 2 to 5% HNO3 | |
| Miscible with water | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Co+2] | |
| 182.941 | |
| CHEBI:86209 | |
| AAS |
| 1 mg/ml Co in 2% HNO3 | |
| 10141-05-6 , 7732-18-5 | |
| CoN2O6 | |
| MFCD00010935 | |
| UFMZWBIQTDUYBN-UHFFFAOYSA-N | |
| cobalt(2+);dinitrate | |
| 25000 | |
| 58.93 |
RUO – Research Use Only