missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Nitrate, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
$306.38 - $22568.12
Chemical Identifiers
| CAS | 10026-22-9 |
|---|---|
| Molecular Formula | CoH12N2O12 |
| Molecular Weight (g/mol) | 291.03 |
| MDL Number | MFCD00149647 |
| InChI Key | QGUAJWGNOXCYJF-UHFFFAOYSA-N |
| IUPAC Name | λ²-cobalt(2+) hexahydrate dinitrate |
| SMILES | O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18609846
|
Spectrum Chemical Mfg Cor
C131525GM |
25 g | Amber Glass Bottle |
Each for $306.38
|
|
||||
|
18609847
|
Spectrum Chemical Mfg Cor
C1315125GM |
125 g | Amber Glass Bottle |
Each for $602.59
|
|
||||
|
18609848
|
Spectrum Chemical Mfg Cor
C1315500GM |
500 g | Amber Glass Bottle |
Each for $1,383.97
|
|
||||
|
18609849
|
Spectrum Chemical Mfg Cor
C131525KG |
2.5 kg | Amber Glass Bottle |
Each for $4,036.74
|
|
||||
|
18609850
|
Spectrum Chemical Mfg Cor
C131512KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18609851
|
Spectrum Chemical Mfg Cor
C131545KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 10026-22-9 | |
| 291.03 | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| CoH12N2O12 | |
| MFCD00149647 | |
| λ²-cobalt(2+) hexahydrate dinitrate |
Specifications
| 10026-22-9 | |
| 25 g | |
| MFCD00149647 | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 291.03 | |
| ACS |
| 1 | |
| CoH12N2O12 | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| λ²-cobalt(2+) hexahydrate dinitrate | |
| 98 to 102% | |
| Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.