missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Nitrate, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Percent Purity | 98 to 102% |
| Grade | ACS |
| Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.