missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Nitrate, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C131512KG
Specifications
| 10026-22-9 | |
| 12 kg | |
| MFCD00149647 | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 291.03 | |
| ACS |
| 1 | |
| CoH12N2O12 | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| λ²-cobalt(2+) hexahydrate dinitrate | |
| 98 to 102% | |
| Poly Pail |
Chemical Identifiers
| 10026-22-9 | |
| 291.03 | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| CoH12N2O12 | |
| MFCD00149647 | |
| λ²-cobalt(2+) hexahydrate dinitrate |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.