missing translation for 'onlineSavingsMsg'
Learn More

Cobalt ICP Standard, 1000 ppm Co in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PCO1KN500

 View more versions of this product

Catalog No. PCO1KN-500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cobalt ICP Standard
7440-48-4 , 7697-37-2 , 7440-48-4
93.0,4.89,0.09
1 mL = 1 mg Co
Co
Pink
Approximately 0°C
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
500 mL
CHEBI:27638
Natural Poly Bottle
1,000ppm Co
97.0,5.09,0.1
Liquid
MFCD00010935
< 2
Standard
[Co]
58.93
104730

Chemical Identifiers

7440-48-4
58.93
GUTLYIVDDKVIGB-UHFFFAOYSA-N
CHEBI:27638
[Co]
Co
MFCD00010935
104730
cobalt
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions