missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt ICP Standard, 1000 ppm Co in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PCO1KN100
Specifications
| Cobalt ICP Standard | |
| 7440-48-4 , 7697-37-2 , 7440-48-4 | |
| 93.0,4.89,0.09 | |
| 1 mL = 1 mg Co | |
| Co | |
| Pink | |
| Approximately 0°C | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| cobalt | |
| 100 mL | |
| CHEBI:27638 |
| Natural LDPE Bottle | |
| 1,000ppm Co | |
| 97.0,5.09,0.1 | |
| Liquid | |
| MFCD00010935 | |
| < 2 | |
| Standard | |
| [Co] | |
| 58.93 | |
| 104730 |
Chemical Identifiers
| 7440-48-4 | |
| 58.93 | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| CHEBI:27638 | |
| [Co] |
| Co | |
| MFCD00010935 | |
| 104730 | |
| cobalt |