missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Chloride, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
$248.03 - $26773.44
Chemical Identifiers
| CAS | 7791-13-1 |
|---|---|
| Molecular Formula | Cl2CoH12O6 |
| Molecular Weight (g/mol) | 237.92 |
| MDL Number | MFCD00149652 |
| InChI Key | GFHNAMRJFCEERV-UHFFFAOYSA-L |
| IUPAC Name | λ²-cobalt(2+) hexahydrate dichloride |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18609836
|
Spectrum Chemical Mfg Cor
C130525GM |
25 g | Amber Glass Bottle |
Each for $248.03
|
|
||||
|
18609837
|
Spectrum Chemical Mfg Cor
C1305125GM |
125 g | Amber Glass Bottle |
Each for $550.06
|
|
||||
|
18609838
|
Spectrum Chemical Mfg Cor
C1305500GM |
500 g | Amber Glass Bottle |
Each for $1,171.59
|
|
||||
|
18609839
|
Spectrum Chemical Mfg Cor
C130525KG |
2.5 kg | Amber Glass Bottle |
Each for $4,800.21
|
|
||||
|
18609840
|
Spectrum Chemical Mfg Cor
C130512KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18609841
|
Spectrum Chemical Mfg Cor
C130550KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7791-13-1 | |
| 237.92 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
| Cl2CoH12O6 | |
| MFCD00149652 | |
| λ²-cobalt(2+) hexahydrate dichloride |
Specifications
| 7791-13-1 | |
| 25 g | |
| MFCD00149652 | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] | |
| 237.92 | |
| ACS |
| 1 | |
| Cl2CoH12O6 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| λ²-cobalt(2+) hexahydrate dichloride | |
| 98 to 102% | |
| Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.