missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Chloride, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C1305500GM
Specifications
| 7791-13-1 | |
| 500 g | |
| MFCD00149652 | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] | |
| 237.92 | |
| ACS |
| 1 | |
| Cl2CoH12O6 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| λ²-cobalt(2+) hexahydrate dichloride | |
| 98 to 102% | |
| Amber Glass Bottle |
Chemical Identifiers
| 7791-13-1 | |
| 237.92 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
| Cl2CoH12O6 | |
| MFCD00149652 | |
| λ²-cobalt(2+) hexahydrate dichloride |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.