Learn More
Cobalt AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Co), LabChem™
$58.89 - $148.29
Chemical Identifiers
| CAS | 10026-22-9 |
|---|---|
| Molecular Formula | CoH12N2O12 |
| Molecular Weight (g/mol) | 291.03 |
| MDL Number | MFCD00149647 |
| InChI Key | QGUAJWGNOXCYJF-UHFFFAOYSA-N |
| PubChem CID | 24821 |
| ChEBI | CHEBI:86214 |
| IUPAC Name | λ²-cobalt(2+) hexahydrate dinitrate |
| SMILES | O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Chemical Identifiers
| 10026-22-9 | |
| 291.03 | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| CHEBI:86214 | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| CoH12N2O12 | |
| MFCD00149647 | |
| 24821 | |
| λ²-cobalt(2+) hexahydrate dinitrate |
Specifications
| Poly Bottle | |
| 97.51,2,0.49 | |
| 1g/mL | |
| CoH12N2O12 | |
| Co(NO3)2·6H2O | |
| Pink | |
| QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
| λ²-cobalt(2+) hexahydrate dinitrate | |
| 125 mL | |
| CHEBI:86214 | |
| Certified |
| 10026-22-9 , 7697-37-2 , 10026-22-9 | |
| Passes Test | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg Co) | |
| MFCD00149647 | |
| Soluble in water | |
| O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 291.03 | |
| 24821 | |
| 291.03 | |
| Cobalt AA Standard, 1000ppm (1mL = 1mg Co) |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature