missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Co-factors

Catalog No. FERF5101
Change view
Click to view available options
Quantity:
250 μL
800 μL
Product Type:
Methyl Group Donor
Polymerase Co-factor
Concentration:
5 mM
50 mM
100 mM
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Concentration
FERF5101 250 μL Polymerase Co-factor 50 mM
FERC24341 250 μL Polymerase Co-factor 100 mM
FERB13880C 800 μL Methyl Group Donor 5 mM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FERF5101 Supplier Thermo Scientific™ Supplier No. F5101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells

Specifications

Chemical Name or Material Magnesium Chloride
Content And Storage 50 mM MgCl2, 250 μL

Store at -20°C.
Concentration 50 mM
Format Vial
Product Type Polymerase Co-factor
Quantity 250 μL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.