missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat CNTF ELISA Kit

ELISA

$954.00

Specifications

Accession Number P20294
Assay Range 8.23 to 2000 pg/mL
Assay Sensitivity 10 pg/ml
Calibration Range 10 pg/mL to 2000 pg/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ERCNTF
Encompass
View Documents Promotion Details
Invitrogen™
ERCNTF
96 Tests
Pack for $954.00
Only null left
 
Description

Description

The Rat Ciliary NeuroTrophic Factor (Rt CNTF) ELISA quantitates Rt CNTF in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt CNTF. Principle of the method The Rat CNTF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.
Specifications

Specifications

P20294
10 pg/ml
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
96 assays
Colorimetric Microplate Reader
Cntf
<10
HRP
Oncology
2°C to 8°C
1 hrs 20 mins
8.23 to 2000 pg/mL
10 pg/mL to 2000 pg/mL
HRP
Growth Factors & Receptors
This ELISA kit shows no cross-reactivity with any of the cytokines tested: rat CINC-2, CINC-3, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP- 1, TNF-alpha, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
25707
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
CNTF
4 hrs 45 mins
Videos
SDS
Documents

Documents