missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CMA
Description
CMA is a STING agonist; murine-selective (Kd = 3.5 μM). Triggers type I IFN response in murine macrophages, with no effect on human STING signaling.
Specifications
Specifications
| Chemical Name or Material | 9-Oxo-10(9H)-acridineacetic acid |
| CAS | 38609-97-1 |
| Synonym | ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173 |
| Quantity | 10 mg |
| Target | STING Agonists |
| Molecular Formula | C15H11NO3 |
| Formula Weight | 253.25 |
| Purity | 98% |