missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CLS-Y
Used for the isolation of genomic DNA from yeast, algae and fungi. MP Biomedicals CLS-Y is a cell lysis solution. It is a component included in the FastDNA and FastDNA SPIN Kits.
Supplier: MP Biomedicals™ 116540411
Description
- Applications: CLS-Y is a cell lysis solution used for the isolation of genomic DNA from yeast, algae and fungi
- Product Overview: CLS-Y is a component included in the FastDNA and FastDNA SPIN Kits and is a cell lysis solution used for the isolation of genomic DNA from yeast, algae and fungi
- Product Description: CLS-Y is a cell lysis solution used for the isolation of genomic DNA from yeast, algae and fungi
- Recommended Use: FastPrep™ Instrument
Specifications
| Store at room temperature. | |
| Algae, Bacteria (Gram +/-), Fungi, Spores, Yeast | |
| 110 mL | |
| Yeast and Fungi lysis buffer |
| Cell Lysis Solution for Yeast and Fungi (CLS-Y) | |
| Isolation of genomic DNA from yeast, algae and fungi | |
| Up to 500 mg per prep | |
| Liquid |