missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Clobetasol propionate

Catalog No. AC466820010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466820010 1 g
AC466820050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466820010 Supplier Thermo Scientific Chemicals Supplier No. 466820010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25122-46-7
Molecular Formula C25H32ClFO5

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: insoluble.
Melting Point 191.0°C to 195.0°C
Formula Weight 466.97
InChI Key CBGUOGMQLZIXBE-XGQKBEPLSA-N
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl
IUPAC Name (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-1-(2-chloroacetyl)-9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl propanoate
Molecular Weight (g/mol) 466.97
Chemical Name or Material Cloβsol propionate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May damage fertility or the unborn child.

May cause damage to organs through prolonged or repeated exposure.

May cause long lasting harmful effects to aquatic life.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 246-634-3

RUO – Research Use Only