missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin Phosphate USP , Medisca™

$113.46 - $1364.75

Chemical Identifiers

CAS 24729-96-2
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
18-048-023
View Documents
Medisca Pharmaceutique Inc
000601
10 g
Each for $190.15
Only null left
 
18-048-105
View Documents
Medisca Pharmaceutique Inc
000604
25 g
Each for $404.84
Only null left
 
18-048-122
View Documents
Medisca Pharmaceutique Inc
000603
5 g
Each for $113.46
Only null left
 
18-048-129
View Documents
Medisca Pharmaceutique Inc
000605
100 g
Each for $1,364.75
Only null left
 
Description

Description

Clindamycin Phosphate USP is a white to off-white, hygroscopic, crystalline powder. It is odorless to practically odorless.

Specifications

Chemical Identifiers

24729-96-2

Specifications

α-Carboxybenzyl Penicillin Disodium Salt
24729-96-2
10 g
USP
Crystalline Powder
White to Off-White
≥ 758 μg/mg
Videos
SDS
Documents

Documents