missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin Phosphate USP , Medisca™
GREENER_CHOICE

Catalog No. 18048023
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048023 10 g
18048105 25 g
18048122 5 g
18048129 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18048023 Supplier Medisca Pharmaceutique Inc Supplier No. 000601

Clindamycin Phosphate USP is a white to off-white, hygroscopic, crystalline powder. It is odorless to practically odorless.

Chemical Identifiers

CAS 24729-96-2

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White to Off-White
Quantity 10 g
Percent Purity ≥ 758 μg/mg
Grade USP