missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Climbazole 98.0+%, TCI America™
$111.84 - $900.33
Chemical Identifiers
| CAS | 38083-17-9 |
|---|---|
| Molecular Formula | C15H17ClN2O2 |
| Molecular Weight (g/mol) | 292.763 |
| InChI Key | OWEGWHBOCFMBLP-UHFFFAOYSA-N |
| Synonym | 1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone |
| PubChem CID | 37907 |
| ChEBI | CHEBI:83719 |
| IUPAC Name | 1-(4-chlorophenoxy)-1-imidazol-1-yl-3,3-dimethylbutan-2-one |
| SMILES | CC(C)(C)C(=O)C(N1C=CN=C1)OC2=CC=C(C=C2)Cl |
Chemical Identifiers
| 38083-17-9 | |
| 292.763 | |
| 1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone | |
| CHEBI:83719 | |
| CC(C)(C)C(=O)C(N1C=CN=C1)OC2=CC=C(C=C2)Cl |
| C15H17ClN2O2 | |
| OWEGWHBOCFMBLP-UHFFFAOYSA-N | |
| 37907 | |
| 1-(4-chlorophenoxy)-1-imidazol-1-yl-3,3-dimethylbutan-2-one |
Specifications
| 38083-17-9 | |
| White | |
| 25 g | |
| OWEGWHBOCFMBLP-UHFFFAOYSA-N | |
| 1-(4-chlorophenoxy)-1-imidazol-1-yl-3,3-dimethylbutan-2-one | |
| 37907 | |
| 292.76 | |
| Crystalline Powder |
| 98°C | |
| C15H17ClN2O2 | |
| 1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone | |
| CC(C)(C)C(=O)C(N1C=CN=C1)OC2=CC=C(C=C2)Cl | |
| 292.763 | |
| CHEBI:83719 | |
| ≥98.0% (HPLC,T) | |
| Climbazole |
Safety and Handling
RTECSNumber : EL7085000
TSCA : No