missing translation for 'onlineSavingsMsg'
Learn More

Climbazole 98.0+%, TCI America™

Supplier:  TCI America C2025500G

 View more versions of this product

Catalog No. C2025500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Climbazole
98°C
C15H17ClN2O2
1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone
CC(C)(C)C(=O)C(N1C=CN=C1)OC2=CC=C(C=C2)Cl
292.763
CHEBI:83719
≥98.0% (HPLC,T)
38083-17-9
White
500 g
OWEGWHBOCFMBLP-UHFFFAOYSA-N
1-(4-chlorophenoxy)-1-imidazol-1-yl-3,3-dimethylbutan-2-one
37907
292.76
Crystalline Powder

Chemical Identifiers

38083-17-9
292.763
1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethyl-2-butanone
CHEBI:83719
CC(C)(C)C(=O)C(N1C=CN=C1)OC2=CC=C(C=C2)Cl
C15H17ClN2O2
OWEGWHBOCFMBLP-UHFFFAOYSA-N
37907
1-(4-chlorophenoxy)-1-imidazol-1-yl-3,3-dimethylbutan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : EL7085000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions