missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clear Vials for Waters WISP™ 96 Position Autosampler, Sold by MilliporeSigma™ Supelco™
Available in a variety of volumes and closure types. Vials feature thicker walls for safer sample handling.
| Catalog Number | Mfr. No. | Volume (Metric) | Includes | No. per Pack | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Includes | No. per Pack | Price | Quantity & Availability | |||
|
111011852
|
N/A | 0.7 mL | Snap Plug | 100/Pk. |
N/A
|
|
|||
|
111011812
|
N/A | 1 mL | Snap Plug | 200/Pk. |
N/A
|
|
|||
|
111010282
|
N/A | 1 mL | Snap Plug | 1000/Pk. |
N/A
|
|
|||
|
111012026
|
N/A | 1 mL | Vial Only | 200/Pk. |
N/A
|
|
|||
Specifications
| Waters WISP™ 96 Position Autosampler |
Waters is a registered trademark of Waters