Learn More
Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates

Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.
$597.52 - $1,341.94
Specifications
| Barcode | No Barcode |
|---|---|
| Format | Standard |
| Detection Method | Colorimetric |
| Array | 8 x 12 |
| Color | Clear |
Alert:
Each lot is functionally tested for IgG binding.
| Catalog Number | Mfr. No. | Surface Treatment | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Surface Treatment | Price | Quantity & Availability | |||||
1424577
![]() |
Thermo Scientific™
3555 |
Immulon 1 B |
Case of 50 Each for $597.52
|
|
|||||
1424579
![]() |
Thermo Scientific™
3655 |
Immulon 2 HB |
Case of 50 Each for $661.39
|
|
|||||
|
12565137
|
Thermo Scientific™
449824 |
MaxiSorp |
Case of 60 Each for $1,341.94
|
|
|||||
|
12565139
|
Thermo Scientific™
475434 |
PolySorp |
Case of 60 Each for $1,317.53
|
|
|||||
Description
Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
- Clear polystyrene
- Choice of a variety of surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific PolySorp and Immulon 1B are for adsorption of hydrophobic molecules.
- Thermo Scientific Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
Ordering Information:
Please note that availability may vary by country.
Specifications
| No Barcode | |
| Colorimetric | |
| Clear | |
| Polystyrene | |
| Non-sterile | |
| None |
| Standard | |
| 8 x 12 | |
| Microplate | |
| 96 | |
| Round |