missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates
GREENER_CHOICE

Catalog No. 1424579
Change view
Click to view available options
Surface Treatment:
Immulon 1 B
Immulon 2 HB
MaxiSorp
PolySorp
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
1424579 Immulon 2 HB
1424577 Immulon 1 B
12565137 MaxiSorp
12565139 PolySorp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Each lot is functionally tested for IgG binding.
Catalog No. 1424579 Supplier Thermo Scientific™ Supplier No. 3655
Each lot is functionally tested for IgG binding.

Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.

Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces

  • PolySorp and Immulon 1B for adsorption of hydrophobic molecules
  • Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules
  • MaxiSorp is hydrophilic and ideal for antibody sandwich assays

Ordering Information:

Please note that availability may vary by country.

Specifications

Barcode No Barcode
Binding Property Slightly Hydrophilic
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 50 Cases
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 280 μL
No. of Wells 96
Volume (Metric) Working 180 μL
Sterility Non-sterile
Surface Treatment Immulon 2 HB
Well Shape Round
No. per Case 50
No. per Pack 10
Plate Blocking None
Target Molecule Proteins
Show More Show Less