Learn More
New Pig CleanMax™ Cleanroom Hood
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this sterile/hood with a bound edge around the open face
$1102.41 - $1200.41
| Catalog Number | Mfr. No. | Sterility | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sterility | Price | Quantity | |||||
|
17-987-943
|
New Pig
WPL3009 |
Non-sterile |
Case of 100 Each for $1,102.41
|
|
|||||
|
17-987-944
|
New Pig
WPL3010 |
Sterile |
Case of 100 Each for $1,200.41
|
|
|||||
Description
- Sterile for Critical Environments: This hood is sterile, making it suitable for high-purity applications where protection from pathogens and viruses is essential.
- Effective Barrier Protection: It provides an excellent barrier against dry particulates, microorganisms, and non-hazardous liquids, safeguarding both the wearer and the cleanroom environment.
- Low-Linting and Low-Particulate: The material is designed to shed minimal lint and particles, a critical feature for maintaining the integrity of controlled environments.
- Secure and Comfortable Fit: The bound edge around the open face helps to create a secure seal while still providing a comfortable fit for the wearer.
- Ideal for Aseptic Gowning: As a key component of a full cleanroom gowning ensemble, it helps to ensure head and neck protection during aseptic procedures.
- Durable Construction: Made from a composite fabric, the hood offers enhanced strength and resistance to tears, ensuring reliable performance during use.
Specifications
| Sterile |