Learn More
New Pig CleanMax™ Cleanroom Hood
Catalog No. 17987943
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
17987943 Non-sterile
17987944 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17987943 Supplier New Pig Supplier No. WPL3009
Only null left
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this sterile/hood with a bound edge around the open face
  • Sterile for Critical Environments: This hood is sterile, making it suitable for high-purity applications where protection from pathogens and viruses is essential.
  • Effective Barrier Protection: It provides an excellent barrier against dry particulates, microorganisms, and non-hazardous liquids, safeguarding both the wearer and the cleanroom environment.
  • Low-Linting and Low-Particulate: The material is designed to shed minimal lint and particles, a critical feature for maintaining the integrity of controlled environments.
  • Secure and Comfortable Fit: The bound edge around the open face helps to create a secure seal while still providing a comfortable fit for the wearer.
  • Ideal for Aseptic Gowning: As a key component of a full cleanroom gowning ensemble, it helps to ensure head and neck protection during aseptic procedures.
  • Durable Construction: Made from a composite fabric, the hood offers enhanced strength and resistance to tears, ensuring reliable performance during use.
Specifications
Sterility Non-sterile
Style Sterile