Learn More
New Pig CleanMax™ Cleanroom Hood
Description
- Sterile for Critical Environments: This hood is sterile, making it suitable for high-purity applications where protection from pathogens and viruses is essential.
- Effective Barrier Protection: It provides an excellent barrier against dry particulates, microorganisms, and non-hazardous liquids, safeguarding both the wearer and the cleanroom environment.
- Low-Linting and Low-Particulate: The material is designed to shed minimal lint and particles, a critical feature for maintaining the integrity of controlled environments.
- Secure and Comfortable Fit: The bound edge around the open face helps to create a secure seal while still providing a comfortable fit for the wearer.
- Ideal for Aseptic Gowning: As a key component of a full cleanroom gowning ensemble, it helps to ensure head and neck protection during aseptic procedures.
- Durable Construction: Made from a composite fabric, the hood offers enhanced strength and resistance to tears, ensuring reliable performance during use.
Specifications
Specifications
| Sterility | Sterile |
| Style | Sterile |